Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031827.2 | NP_001026997.1 | 3 | 969 | ||
NM_001320579.1 | NP_001307508.1 | 3 | 931 | |||
NR_135304.1 | 2 | 287 | ||||
GenBank mRNA | AF060511.2 | - | 3 | 358 | ||
AK307153.1 | - | 2 | 644 | |||
BC022832.1 | - | 3 | 969 | 4647797 | ||
BC144688.1 | - | 3 | 656 | 9053222 | ||
BQ639539.1 | - | 2 | 264 | |||
BU590412.1 | - | 3 | 226 |
Gene Symbol: BOLA2-SMG1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027255.1 | - | 2 | 645 |
Gene Symbol: BOLA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039182.2 | NP_001034271.1 | 3 | 969 | ||
NM_001320607.1 | NP_001307536.1 | 3 | 931 | |||
GenBank mRNA | BC137532.1 | - | 3 | 656 | 9021532 | |
BC137534.1 | - | 3 | 656 | 9021534 | ||
BU960186.1 | - | 3 | 294 |