Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268547.3 | XP_005268604.1 | 1 | 24 | ||
XM_005268548.3 | XP_005268605.1 | 1 | 24 | |||
XM_005268552.2 | XP_005268609.1 | 1 | 24 | |||
XM_011537715.1 | XP_011536017.1 | 1 | 24 | |||
XM_011537716.2 | XP_011536018.1 | 1 | 24 |