Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NXF2 SIRNA Assays ›
          Gene Symbol:
          NXF2 NXF2B
          Gene Name:
          nuclear RNA export factor 2
          nuclear RNA export factor 2B
          Gene Aliases:
          CT39, TAPL-2, TCP11X2
          TCP11X1, bA353J17.1
          Chromosome Location:
          Chr.X: 102247100 - 102326720 on Build GRCh38
          Chromosome Location:
          Chr.X: 102360395 - 102440008 on Build GRCh38
          UniGene ID:
          Hs.530358
          Hs.530358
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s198498

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NXF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022053.3 NP_071336.1 12 1333
          GenBank mRNA AF285596.1 - 9 932
          AJ277659.1 - 16 1819
          AK000423.1 - 8 729
          AK308255.1 - 12 1272
          BC015020.1 - 12 1253 3921074
          Gene Symbol: NXF2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099686.2 NP_001093156.1 12 1333

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA export from nucleus
          multicellular organism development
          poly(A)+ mRNA export from nucleus
          RNA transport
          nucleotide binding
          RNA binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0