Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other USP17L9P SIRNA Assays ›
          Gene Symbol:
          USP17L9P USP17L11 USP17L12 USP17L15 USP17L17 USP17L18 USP17L19 USP17L20 USP17L21 USP17L22 USP17L24 USP17L25 USP17L26 USP17L27 USP17L28 USP17L29 USP17L30 USP17L5 USP17L6P
          Gene Name:
          ubiquitin specific peptidase 17-like family member 9, pseudogene
          ubiquitin specific peptidase 17-like family member 11
          ubiquitin specific peptidase 17-like family member 12
          ubiquitin specific peptidase 17-like family member 15
          ubiquitin specific peptidase 17-like family member 17
          ubiquitin specific peptidase 17-like family member 18
          ubiquitin specific peptidase 17-like family member 19
          ubiquitin specific peptidase 17-like family member 20
          ubiquitin specific peptidase 17-like family member 21
          ubiquitin specific peptidase 17-like family member 22
          ubiquitin specific peptidase 17-like family member 24
          ubiquitin specific peptidase 17-like family member 25
          ubiquitin specific peptidase 17-like family member 26
          ubiquitin specific peptidase 17-like family member 27
          ubiquitin specific peptidase 17-like family member 28
          ubiquitin specific peptidase 17-like family member 29
          ubiquitin specific peptidase 17-like family member 30
          ubiquitin specific peptidase 17-like family member 5
          ubiquitin specific peptidase 17-like family member 6, pseudogene
          Gene Aliases:
          RS447, USP17, USP17A, USP17H, USP17I, USP17J, USP17K, USP17L, USP17M
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          DUB4, USP17L6, vDUB4
          Chromosome Location:
          Chr.4: 9358382 - 9359974 on Build GRCh38
          Chromosome Location:
          Chr.4: 9215405 - 9216997 on Build GRCh38
          Chromosome Location:
          Chr.4: 9220152 - 9221744 on Build GRCh38
          Chromosome Location:
          Chr.4: 9234385 - 9236334 on Build GRCh38
          Chromosome Location:
          Chr.4: 9243879 - 9245471 on Build GRCh38
          Chromosome Location:
          Chr.4: 9248630 - 9250222 on Build GRCh38
          Chromosome Location:
          Chr.4: 9253378 - 9254970 on Build GRCh38
          Chromosome Location:
          Chr.4: 9258124 - 9259716 on Build GRCh38
          Chromosome Location:
          Chr.4: 9262872 - 9264464 on Build GRCh38
          Chromosome Location:
          Chr.4: 9267619 - 9269211 on Build GRCh38
          Chromosome Location:
          Chr.4: 9325165 - 9326757 on Build GRCh38
          Chromosome Location:
          Chr.4: 9329911 - 9331503 on Build GRCh38
          Chromosome Location:
          Chr.4: 9334658 - 9336250 on Build GRCh38
          Chromosome Location:
          Chr.4: 9344148 - 9345740 on Build GRCh38
          Chromosome Location:
          Chr.4: 9348893 - 9350485 on Build GRCh38
          Chromosome Location:
          Chr.4: 9353638 - 9355230 on Build GRCh38
          Chromosome Location:
          Chr.4: 9363129 - 9364721 on Build GRCh38
          Chromosome Location:
          Chr.4: 9339403 - 9340995 on Build GRCh38
          Chromosome Location:
          Chr.4: 9367874 - 9369070 on Build GRCh38
          UniGene ID:
          Hs.553810
          Hs.722783
          Hs.741128
          Hs.528621
          Hs.741129
          Hs.741138
          Hs.741137
          Hs.722783
          Hs.741136
          Hs.723053
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.740243
          Hs.722658
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s198523

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: USP17L9P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046416.1 1 69
          GenBank mRNA BC100991.2 - 1 68
          BC100992.2 - 1 68
          BC100993.1 - 1 71
          Gene Symbol: USP17L11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256854.1 NP_001243783.1 1 68
          Gene Symbol: USP17L12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256853.1 NP_001243782.1 1 68
          Gene Symbol: USP17L15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256894.1 NP_001243823.1 1 68
          Gene Symbol: USP17L17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256857.1 NP_001243786.1 1 68
          Gene Symbol: USP17L18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256859.1 NP_001243788.1 1 68
          Gene Symbol: USP17L19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256860.1 NP_001243789.1 1 68
          Gene Symbol: USP17L20
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256861.1 NP_001243790.1 1 68
          Gene Symbol: USP17L21
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256862.1 NP_001243791.1 1 68
          Gene Symbol: USP17L22
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256863.1 NP_001243792.1 1 68
          Gene Symbol: USP17L24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242327.1 NP_001229256.1 1 68
          Gene Symbol: USP17L25
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242326.1 NP_001229255.1 1 68
          Gene Symbol: USP17L26
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242328.1 NP_001229257.1 1 68
          Gene Symbol: USP17L27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242330.1 NP_001229259.1 1 68
          Gene Symbol: USP17L28
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242331.1 NP_001229260.1 1 68
          Gene Symbol: USP17L29
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242332.1 NP_001229261.1 1 68
          Gene Symbol: USP17L30
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256867.1 NP_001243796.1 1 68
          Gene Symbol: USP17L5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242329.1 NP_001229258.1 1 68
          Gene Symbol: USP17L6P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027279.1 1 68

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          apoptotic process
          protein deubiquitination
          thiol-dependent ubiquitinyl hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0