Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALG3 SIRNA Assays ›
          Gene Symbol:
          ALG3
          Gene Name:
          ALG3, alpha-1,3- mannosyltransferase
          Gene Aliases:
          CDG1D, CDGS4, CDGS6, D16Ertd36e, NOT56L, Not56, not
          Chromosome Location:
          Chr.3: 184242329 - 184249525 on Build GRCh38
          UniGene ID:
          Hs.478481
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19865
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALG3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006941.2 NP_001006942.1 2 358
          NM_005787.5 NP_005778.1 2 285
          NR_024534.1 2 248
          XM_011512322.1 XP_011510624.1 2 170
          XM_011512323.2 XP_011510625.1 2 156
          GenBank mRNA AK289361.1 - 2 314
          BC002839.2 - 2 260 3636604
          BC004313.1 - 2 240
          BC021062.1 - 2 249
          BM920198.1 - 2 214
          BX339649.2 - 2 363
          KP189279.1 - 2 278
          KP189280.1 - 2 241
          KP189282.1 - 2 395
          KP189284.1 - 2 276
          KP189285.1 - 2 239
          KP189287.1 - 2 194
          KP189289.1 - 2 275
          KP189290.1 - 2 239
          KP189292.1 - 2 194
          KP189294.1 - 2 254
          KP189295.1 - 2 254
          KP189296.1 - 2 217
          KP189297.1 - 2 217
          KP189299.1 - 2 110
          KP189301.1 - 2 217
          KP189302.1 - 2 275
          KP189303.1 - 2 237
          KP189305.1 - 2 194
          KP189307.1 - 2 274
          KP189308.1 - 2 237
          KP189310.1 - 2 194
          KP189312.1 - 2 274
          KP189313.1 - 2 274
          KP189314.1 - 2 274
          KP189315.1 - 2 237
          KP189316.1 - 2 237
          KP189317.1 - 2 237
          KP189321.1 - 2 194
          KP189322.1 - 2 194
          KP189323.1 - 2 194
          Y09022.1 - 2 285

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00271780_m1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          dolichol-linked oligosaccharide biosynthetic process
          mannosylation
          alpha-1,3-mannosyltransferase activity
          dol-P-Man:Man(5)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline