Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276965.4 | XP_005277022.1 | 5 | 788 | ||
XM_006712884.3 | XP_006712947.1 | 5 | 1212 | |||
XM_006712887.3 | XP_006712950.1 | 7 | 1165 | |||
XM_017005467.1 | XP_016860956.1 | 4 | 784 | |||
XM_017005468.1 | XP_016860957.1 | 4 | 692 | |||
XM_017005469.1 | XP_016860958.1 | 4 | 681 |
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055744.1 | - | 1 | 1239 |