Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GPHN SIRNA Assays ›
Gene Symbol:
GPHN
Gene Name:
gephyrin
Gene Aliases:
GEPH, GPH, GPHRYN, HKPX1, MOCODC
Chromosome Location:
Chr.14: 66507407 - 67735831 on Build GRCh38
UniGene ID:
Hs.208765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20002
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GPHN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024218.1
NP_001019389.1
15
2605
NM_020806.4
NP_065857.1
16
2704
XM_005267254.3
XP_005267311.1
16
1957
XM_011536340.2
XP_011534642.1
19
2345
XM_011536342.2
XP_011534644.1
18
2306
XM_011536343.2
XP_011534645.1
18
2273
XM_011536344.2
XP_011534646.1
18
2246
XM_011536345.2
XP_011534647.1
17
2216
XM_011536346.2
XP_011534648.1
17
2008
XM_011536347.2
XP_011534649.1
16
1603
XM_017020913.1
XP_016876402.1
19
2336
XM_017020914.1
XP_016876403.1
18
2288
XM_017020915.1
XP_016876404.1
18
2237
XM_017020916.1
XP_016876405.1
17
2198
XM_017020917.1
XP_016876406.1
17
2189
XM_017020918.1
XP_016876407.1
17
2174
XM_017020919.1
XP_016876408.1
16
2150
XM_017020920.1
XP_016876409.1
17
2139
XM_017020921.1
XP_016876410.1
19
2408
XM_017020922.1
XP_016876411.1
20
1874
XM_017020923.1
XP_016876412.1
17
2279
XM_017020924.1
XP_016876413.1
19
2339
XM_017020926.1
XP_016876415.1
18
2267
GenBank mRNA
AB037806.1
-
15
2605
AF272663.1
-
15
2605
AJ272033.1
-
15
1484
AJ272343.1
-
16
1623
AK294532.1
-
18
1646
AK295577.1
-
14
1481
AK297447.1
-
14
1406
AK315569.1
-
16
1953
BC030016.2
-
16
1595
4826821
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Mo-molybdopterin cofactor biosynthetic process
molybdenum incorporation into molybdenum-molybdopterin complex
molybdopterin cofactor biosynthetic process
glycine receptor clustering
gamma-aminobutyric acid receptor clustering
protein binding
ATP binding
metal ion binding
molybdopterin adenylyltransferase activity
molybdopterin molybdotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0