Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD20A5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040113.1 | 4 | 875 | |||
GenBank mRNA | AK092114.1 | - | 3 | 1120 |
Gene Symbol: ANKRD20A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003366.2 | 4 | 483 |
Gene Symbol: ANKRD20A9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027995.1 | 1 | 87 |