Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHRFAM7A SIRNA Assays ›
          Gene Symbol:
          CHRFAM7A CHRNA7
          Gene Name:
          CHRNA7 (exons 5-10) and FAM7A (exons A-E) fusion
          cholinergic receptor nicotinic alpha 7 subunit
          Gene Aliases:
          CHRNA7, CHRNA7-DR1, D-10
          CHRNA7-2, NACHRA7
          Chromosome Location:
          Chr.15: 30357770 - 30393849 on Build GRCh38
          Chromosome Location:
          Chr.15: 32030462 - 32172521 on Build GRCh38
          UniGene ID:
          Hs.510853
          Hs.511772
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s200391

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHRFAM7A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_139320.1 NP_647536.1 10 1322
          NM_148911.1 NP_683709.1 9 1258
          XM_005254750.2 XP_005254807.1 11 1491
          XM_011522153.1 XP_011520455.1 11 1016
          XM_011522155.2 XP_011520457.1 9 1207
          GenBank mRNA AF036903.1 - 8 768
          AF037646.1 - 7 892
          AK292984.1 - 10 1089
          BC101345.1 - 11 1195 40025185
          BC101346.2 - 10 1021 40025186
          BC101347.2 - 10 1021 40025187
          BC101348.2 - 10 1021 40025188
          Gene Symbol: CHRNA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000746.5 NP_000737.1 10 1138
          NM_001190455.2 NP_001177384.1 10 1225
          NR_046324.1 8 948
          XM_011521176.2 XP_011519478.2 9 993
          XM_011521177.2 XP_011519479.1 7 1078
          XM_011521178.2 XP_011519480.1 9 1028
          XM_017021882.1 XP_016877371.1 11 1203
          XM_017021883.1 XP_016877372.1 11 1200
          XM_017021884.1 XP_016877373.1 9 1049
          GenBank mRNA AF385585.1 - 10 1040
          AJ586911.1 - 10 1043
          AK292069.1 - 10 1108
          AK294229.1 - 10 1220
          BC037571.1 - 8 875 5245812
          EF405809.1 - 7 715
          EF405817.1 - 7 716
          L25827.1 - 9 1003
          U40583.1 - 10 1033
          U62436.1 - 10 1098
          X70297.1 - 10 1129
          Y08420.1 - 10 1026
          Z23141.1 - 9 957

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transmembrane transport
          activation of MAPK activity
          response to hypoxia
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          signal transduction
          synaptic transmission, cholinergic
          memory
          positive regulation of cell proliferation
          negative regulation of tumor necrosis factor production
          response to nicotine
          positive regulation of angiogenesis
          cognition
          cation transmembrane transport
          extracellular ligand-gated ion channel activity
          beta-amyloid binding
          acetylcholine-activated cation-selective channel activity
          ligand-gated ion channel activity
          acetylcholine receptor activity
          toxic substance binding
          chloride channel regulator activity
          acetylcholine-gated cation channel activity
          acetylcholine binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0