siRNA Details
Gene Symbol: Sncb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033610.2 | NP_291088.1 | 6 | 1085 | ||
GenBank mRNA | BC019409.1 | - | 6 | 1085 | 4482283 |

Gene Symbol: Sncb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033610.2 | NP_291088.1 | 6 | 1085 | ||
GenBank mRNA | BC019409.1 | - | 6 | 1085 | 4482283 |