Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145126.2 | NP_660108.2 | 7, 8 | 751 | ||
GenBank mRNA | AK131914.1 | - | 7, 8 | 752 | ||
AY049765.2 | - | 7, 8 | 736 | |||
AY065557.1 | - | 7, 8 | 736 | |||
BC130015.1 | - | 7, 8 | 740 | 40140436 |