Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271416.1 | NP_001258345.1 | 532 | |||
NM_001271417.1 | NP_001258346.1 | 517 | ||||
NM_001271418.1 | NP_001258347.1 | 496 | ||||
NM_001271419.1 | NP_001258348.1 | 486 | ||||
NM_001271446.1 | NP_001258375.1 | 512 | ||||
NM_010738.3 | NP_034868.1 | 501 | ||||
GenBank mRNA | AK076200.1 | - | 363 | |||
BC002070.1 | - | 3 | 358 | 3486025 | ||
BG085582.2 | - | 355 | ||||
BY228083.1 | - | 375 | ||||
EU704682.1 | - | 3 | 346 | |||
J03636.1 | - | 365 | ||||
M18184.1 | - | 3 | 332 |