Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTB SIRNA Assays ›
          Gene Symbol:
          Actb
          Gene Name:
          actin, beta
          Gene Aliases:
          Actx, E430023M04Rik, beta-actin
          Chromosome Location:
          Chr.5: 142903116 - 142906724 on Build GRCm38
          UniGene ID:
          Mm.328431
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s200989

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Actb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB028847.1 - 2 150
          AK075973.1 - 5 942
          AK078935.1 - 5 945
          AK088691.1 - 5 941
          AK145191.1 - 5 944
          AK145196.1 - 5 944
          AK145308.1 - 5 944
          AK147787.1 - 5 941
          AK149920.1 - 5 943
          AK150545.1 - 5 945
          AK150553.1 - 5 945
          AK150558.1 - 5 945
          AK150570.1 - 5 946
          AK150662.1 - 5 943
          AK150675.1 - 5 943
          AK150711.1 - 5 944
          AK150854.1 - 5 945
          AK150866.1 - 5 944
          AK150879.1 - 5 944
          AK151010.1 - 5 941
          AK151136.1 - 5 944
          AK151145.1 - 5 941
          AK151159.1 - 5 944
          AK151166.1 - 5 941
          AK151190.1 - 5 944
          AK151202.1 - 5 944
          AK151226.1 - 5 941
          AK151277.1 - 5 941
          AK151297.1 - 5 944
          AK151350.1 - 5 941
          AK151385.1 - 5 944
          AK151386.1 - 5 941
          AK151398.1 - 5 944
          AK151444.1 - 5 944
          AK151468.1 - 5 941
          AK151663.1 - 5 944
          AK151712.1 - 5 946
          AK151898.1 - 5 942
          AK151956.1 - 5 941
          AK151995.1 - 5 944
          AK151999.1 - 5 944
          AK152043.1 - 5 945
          AK152052.1 - 5 942
          AK152083.1 - 5 945
          AK152218.1 - 5 945
          AK152433.1 - 5 942
          AK152470.1 - 5 942
          AK152591.1 - 5 945
          AK152615.1 - 5 944
          AK152651.1 - 5 944
          AK152671.1 - 5 945
          AK152813.1 - 5 947
          AK152841.1 - 5 945
          AK152844.1 - 5 941
          AK153461.1 - 5 941
          AK159759.1 - 5 944
          AK159834.1 - 5 941
          AK160029.1 - 5 941
          AK166349.1 - 5 944
          AK166498.1 - 5 944
          AK167117.1 - 5 944
          AK167825.1 - 5 944
          AK167960.1 - 5 944
          BC040513.1 - 2 141
          BC138611.1 - 5 909 8861631
          BC138614.1 - 5 909 8861634
          J04181.1 - 5 925
          M12481.1 - 4 785
          U89400.1 - 2 291
          X03672.1 - 5 943
          X03765.1 - 4 785

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031144, FQ232650, FQ228822, FQ229293, FQ226945, and others.

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0