Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243043.1 | NP_001229972.1 | 17, 18 | 2766 | ||
NM_001243044.1 | NP_001229973.1 | 16, 17 | 2548 | |||
NM_007454.3 | NP_031480.2 | 16, 17 | 2548 | |||
GenBank mRNA | AA003413.1 | - | 2, 3 | 184 | ||
AK034140.1 | - | 17, 18 | 2769 | |||
AK155892.1 | - | 16, 17 | 2468 | |||
AK159273.1 | - | 16, 17 | 2439 | |||
AK160042.1 | - | 16, 17 | 2439 | |||
AK165747.1 | - | 16, 17 | 2466 | |||
AK170376.1 | - | 16, 17 | 2459 | |||
AK196462.1 | - | 1, 2 | 87 | |||
BC008513.1 | - | 16, 17 | 2361 | 3601435 | ||
BC066827.1 | - | 7, 8 | 1092 | |||
Y07919.1 | - | 16, 17 | 2403 |