Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010104.3 | NP_034234.1 | 2 | 720 | ||
GenBank mRNA | AB081657.1 | - | 2 | 106 | ||
AK040778.1 | - | 2 | 378 | |||
AK152647.1 | - | 2 | 371 | |||
AK153437.1 | - | 2 | 375 | |||
BC029547.1 | - | 2 | 514 | 5059522 | ||
D43775.1 | - | 2 | 431 | |||
U35233.1 | - | 2 | 125 |