Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008001.4 | NP_032027.2 | 11 | 2391 | ||
GenBank mRNA | AK148151.1 | - | 11 | 2392 | ||
BC004838.1 | - | 3 | 281 | |||
BC011462.1 | - | 11 | 2328 | 3674293 | ||
U22325.1 | - | 11 | 1990 |