Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010358.5 | NP_034488.1 | 5 | 542 | ||
GenBank mRNA | BC003822.1 | - | 5 | 365 | 3600444 | |
BC046758.1 | - | 5 | 395 | 5693552 | ||
BC091763.1 | - | 5 | 375 | 5102093 | ||
J03952.1 | - | 5 | 356 | |||
J04632.1 | - | 5 | 543 |