Search
Search

Idh1| Gene Symbol: Idh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111320.1 | NP_001104790.1 | 3 | 530 | ||
| NM_010497.3 | NP_034627.3 | 3 | 508 | |||
| GenBank mRNA | AF020039.1 | - | 2 | 358 | ||
| AK087063.1 | - | 3 | 410 | |||
| AK149019.1 | - | 3 | 439 | |||
| AK151212.1 | - | 3 | 433 | |||
| AK159173.1 | - | 3 | 439 | |||
| AK160896.1 | - | 3 | 440 | |||
| AK167158.1 | - | 3 | 480 | |||
| AK167583.1 | - | 3 | 444 | |||
| AK168832.1 | - | 3 | 482 | |||
| BC088986.1 | - | 3 | 480 | 6808731 | ||
| CF950262.1 | - | 3 | 469 | |||
| DQ403075.1 | - | 2 | 258 | |||