Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008933.1 | NP_032959.1 | 2 | 503 | ||
GenBank mRNA | AK005729.1 | - | 2 | 512 | ||
BC049612.1 | - | 2 | 508 | 6774072 | ||
M16456.1 | - | 2 | 406 | |||
M27501.1 | - | 2 | 406 | |||
X14004.1 | - | 2 | 503 |