Search
Search

Casc3| Gene Symbol: Casc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138660.2 | NP_619601.2 | 2, 3 | 323 | ||
| GenBank mRNA | AF526276.1 | - | 2, 3 | 297 | ||
| AJ292072.1 | - | 537 | ||||
| AK139608.1 | - | 2, 3 | 362 | |||
| AK145886.1 | - | 2, 3 | 66 | |||
| BC060672.1 | - | 2, 3 | 151 | |||
| BC141296.1 | - | 2, 3 | 436 | 9055908 | ||