Search
Search

Cxcl2| Gene Symbol: Cxcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009140.2 | NP_033166.1 | 4 | 667 | ||
| GenBank mRNA | AK137628.1 | - | 4 | 695 | ||
| AK150450.1 | - | 4 | 575 | |||
| AK155874.1 | - | 3 | 1445 | |||
| AK155916.1 | - | 4 | 694 | |||