Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNAI2 SIRNA Assays ›
          Gene Symbol:
          Snai2
          Gene Name:
          snail family zinc finger 2
          Gene Aliases:
          Slug, Slugh, Snail2
          Chromosome Location:
          Chr.16: 14705859 - 14709385 on Build GRCm38
          UniGene ID:
          Mm.4272
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s202009

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Snai2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011415.2 NP_035545.1 2 417
          GenBank mRNA AK133968.1 - 2 425
          BC062164.1 - 2 417 30242610
          U79550.1 - 2 345
          U97059.1 - 2 302

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013035, U97061, AF497973.

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          epithelial to mesenchymal transition
          epithelial to mesenchymal transition involved in endocardial cushion formation
          cell migration involved in endocardial cushion formation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          negative regulation of cell adhesion involved in substrate-bound cell migration
          Notch signaling pathway
          multicellular organism development
          sensory perception of sound
          response to radiation
          negative regulation of keratinocyte proliferation
          negative regulation of vitamin D biosynthetic process
          neural crest cell development
          cell migration
          positive regulation of cell migration
          negative regulation of chondrocyte differentiation
          regulation of chemokine production
          positive regulation of histone acetylation
          negative regulation of catenin import into nucleus
          desmosome disassembly
          cellular response to platelet-derived growth factor stimulus
          regulation of apoptotic process
          negative regulation of apoptotic process
          pigmentation
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          wound healing, spreading of cells
          positive regulation of fat cell differentiation
          regulation of osteoblast differentiation
          white fat cell differentiation
          palate development
          canonical Wnt signaling pathway
          epithelium development
          cartilage morphogenesis
          regulation of branching involved in salivary gland morphogenesis
          negative regulation of vitamin D receptor signaling pathway
          cellular response to epidermal growth factor stimulus
          cellular response to ionizing radiation
          negative regulation of canonical Wnt signaling pathway
          negative regulation of cell-cell adhesion by negative regulation of transcription from RNA polymerase II promoter
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of stem cell proliferation
          regulation of bicellular tight junction assembly
          negative regulation of anoikis
          positive regulation of endothelial cell chemotaxis
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          nucleic acid binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          sequence-specific DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0