siRNA Details
Gene Symbol: Sox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009233.3 | NP_033259.2 | 1 | 1791 | ||
GenBank mRNA | AB108672.1 | - | 1 | 988 | ||
AI843839.1 | - | 1 | 108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
