Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B3GNT3 SIRNA Assays ›
          Gene Symbol:
          B3GNT3
          Gene Name:
          UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
          Gene Aliases:
          B3GAL-T8, B3GN-T3, B3GNT-3, HP10328, TMEM3, beta3Gn-T3
          Chromosome Location:
          Chr.19: 17794828 - 17813576 on Build GRCh38
          UniGene ID:
          Hs.657825
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s20209

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B3GNT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014256.3 NP_055071.2 3 1805
          XM_011527626.2 XP_011525928.1 3 1764
          GenBank mRNA AB015630.1 - 3 1775
          AB049585.1 - 3 1760
          AK075434.1 - 4 1355
          AK075466.1 - 3 1778
          AK225421.1 - 3 1772
          AY358955.1 - 3 1760

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          O-glycan processing
          keratan sulfate biosynthetic process
          poly-N-acetyllactosamine biosynthetic process
          UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
          N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase activity
          beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline