Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BMP2 SIRNA Assays ›
          Gene Symbol:
          BMP2
          Gene Name:
          bone morphogenetic protein 2
          Gene Aliases:
          BDA2, BMP2A
          Chromosome Location:
          Chr.20: 6767700 - 6780280 on Build GRCh38
          UniGene ID:
          Hs.73853
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2022

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BMP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001200.3 NP_001191.1 3 1294
          XM_011529323.2 XP_011527625.1 2 1339
          GenBank mRNA BC069214.1 - 3 1294
          KC294426.1 - 2 509
          M22489.1 - 3 832

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          skeletal system development
          osteoblast differentiation
          branching involved in ureteric bud morphogenesis
          response to hypoxia
          in utero embryonic development
          epithelial to mesenchymal transition
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          chondrocyte differentiation
          BMP signaling pathway involved in heart induction
          atrioventricular valve morphogenesis
          endocardial cushion morphogenesis
          negative regulation of Wnt signaling pathway involved in heart development
          proteoglycan metabolic process
          regulation of transcription, DNA-templated
          protein phosphorylation
          inflammatory response
          Notch signaling pathway
          cell-cell signaling
          heart development
          negative regulation of cell proliferation
          embryo development
          organ morphogenesis
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of steroid biosynthetic process
          positive regulation of phosphatase activity
          telencephalon development
          telencephalon regionalization
          positive regulation of Wnt signaling pathway
          bone mineralization
          positive regulation of cell migration
          positive regulation of bone mineralization
          BMP signaling pathway
          protein destabilization
          positive regulation of protein binding
          negative regulation of aldosterone biosynthetic process
          positive regulation of osteoblast proliferation
          cardiocyte differentiation
          embryonic heart tube anterior/posterior pattern specification
          bone mineralization involved in bone maturation
          growth
          odontogenesis of dentin-containing tooth
          positive regulation of odontogenesis
          regulation of odontogenesis of dentin-containing tooth
          positive regulation of apoptotic process
          positive regulation of MAPK cascade
          negative regulation of insulin-like growth factor receptor signaling pathway
          cell fate commitment
          positive regulation of fat cell differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of ossification
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of astrocyte differentiation
          mesenchymal cell differentiation
          inner ear development
          negative regulation of calcium-independent cell-cell adhesion
          cardiac muscle cell differentiation
          cardiac muscle tissue morphogenesis
          oxidation-reduction process
          pericardium development
          corticotropin hormone secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          mesenchyme development
          positive regulation of Wnt signaling pathway by BMP signaling pathway
          positive regulation of cartilage development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to organic cyclic compound
          cellular response to BMP stimulus
          mesenchymal cell proliferation involved in ureteric bud development
          negative regulation of canonical Wnt signaling pathway
          positive regulation of p38MAPK cascade
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          negative regulation of cortisol biosynthetic process
          negative regulation of cardiac muscle cell differentiation
          retinol dehydrogenase activity
          receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity
          phosphatase activator activity
          co-receptor binding
          SMAD binding
          protein heterodimerization activity
          BMP receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0