Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153121.2 | NP_694761.1 | 1, 2 | 912 | ||
GenBank mRNA | AK011517.1 | - | 1, 2 | 825 | ||
AK054348.1 | - | 1, 2 | 852 | |||
AK154180.1 | - | 1, 2 | 913 | |||
BC024838.1 | - | 1, 2 | 890 | 5360726 | ||
BC100548.1 | - | 1, 2 | 615 | 5717642 | ||
BC138476.1 | - | 1, 2 | 348 | 8861496 | ||
BC145725.1 | - | 1, 2 | 348 | 40131028 |