Search
Search

Mppe1| Gene Symbol: Mppe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172630.2 | NP_766218.1 | 4 | 975 | ||
| GenBank mRNA | AK041262.1 | - | 4 | 976 | ||
| AK154265.1 | - | 4 | 829 | |||
| AK158446.1 | - | 4 | 698 | |||
| AK172000.1 | - | 4 | 814 | |||
| AK172496.1 | - | 4 | 730 | |||
| BC045146.1 | - | 4 | 601 | 6306245 | ||
| BC086456.1 | - | 4 | 648 | 4949311 | ||