Search
Search

Gdf15| Gene Symbol: Gdf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011819.2 | NP_035949.2 | 1 | 184 | ||
| GenBank mRNA | AF159571.1 | - | 1 | 655 | ||
| AK144199.1 | - | 2 | 483 | |||
| AK159242.1 | - | 1 | 185 | |||
| AK160927.1 | - | 1 | 97 | |||
| BC067248.1 | - | 1 | 173 | 820434 | ||