Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195229.1 | NP_001182158.1 | 26, 27 | 2696 | ||
GenBank mRNA | AK147862.1 | - | 7, 8 | 1080 |