Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007794.1 | NP_001007795.1 | 4 | 826 | ||
NM_006090.3 | NP_006081.1 | 4 | 788 | |||
XM_006710285.3 | XP_006710348.1 | 4 | 780 | |||
XM_011540496.1 | XP_011538798.1 | 4 | 788 | |||
GenBank mRNA | AF068302.1 | - | 4 | 746 | ||
AF138862.1 | - | 3 | 656 | |||
AK023381.1 | - | 4 | 775 | |||
BC032610.2 | - | 5 | 788 | 5554219 | ||
BC049196.1 | - | 4 | 826 | 5226538 |