Search
Search

BASP1| Gene Symbol: BASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271606.1 | NP_001258535.1 | 2 | 1167 | ||
| NM_006317.4 | NP_006308.3 | 2 | 1342 | |||
| GenBank mRNA | AF039656.1 | - | 2 | 1133 | ||
| AK295995.1 | - | 3 | 1108 | |||
| BC000518.2 | - | 2 | 1257 | 2822874 | ||