Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025084.2 | NP_001020255.2 | 2, 3 | 253 | ||
GenBank mRNA | DQ058638.1 | - | 2, 3 | 253 |