Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAPGEF3 SIRNA Assays ›
          Gene Symbol:
          RAPGEF3
          Gene Name:
          Rap guanine nucleotide exchange factor 3
          Gene Aliases:
          CAMP-GEFI, EPAC, EPAC1, HSU79275, bcm910
          Chromosome Location:
          Chr.12: 47734670 - 47759106 on Build GRCh38
          UniGene ID:
          Hs.8578
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s20361

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAPGEF3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098531.2 NP_001092001.1 24, 25 3020
          NM_001098532.2 NP_001092002.1 23, 24 2784
          NM_006105.5 NP_006096.2 24, 25 2554
          XM_005268571.3 XP_005268628.1 23, 24 2530
          XM_011537752.1 XP_011536054.1 24, 25 3037
          XM_011537755.1 XP_011536057.1 24, 25 2599
          XM_011537758.2 XP_011536060.1 24, 25 2649
          GenBank mRNA AF103905.1 - 23, 24 2344
          AK055502.1 - 6, 7 1335
          AK074444.1 - 8, 9 1712
          AK092448.1 - 24, 25 2992
          AK125545.1 - 20, 21 2037
          AK290230.1 - 25, 26 2685
          AK299029.1 - 23, 24 2447
          BC017728.1 - 24, 25 2634 4414360
          BC092404.1 - 24, 25 2623 6181000
          U78168.1 - 24, 25 2559

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_021690, U78167, BC107667.

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          signal transduction
          brain development
          cell proliferation
          negative regulation of cardiac muscle cell apoptotic process
          positive regulation of smooth muscle cell migration
          cAMP-mediated signaling
          spinal cord development
          positive regulation of cAMP catabolic process
          Rap protein signal transduction
          negative regulation of collagen biosynthetic process
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of syncytium formation by plasma membrane fusion
          response to nicotine
          cellular response to drug
          positive regulation of GTPase activity
          positive regulation of angiogenesis
          positive regulation of cell size
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of protein export from nucleus
          regulation of insulin secretion
          positive regulation of stress fiber assembly
          positive regulation of syncytium formation by plasma membrane fusion
          artery development
          establishment of endothelial barrier
          cellular response to cAMP
          cellular response to transforming growth factor beta stimulus
          liver regeneration
          positive regulation of protein acetylation
          positive regulation of GTP binding
          positive regulation of calcium ion transmembrane transport
          positive regulation of hydrogen:potassium-exchanging ATPase activity
          cellular response to L-dopa
          dorsal root ganglion development
          regulation of actin cytoskeleton reorganization
          positive regulation of actin cytoskeleton reorganization
          regulation of histone H3-K9 acetylation
          guanyl-nucleotide exchange factor activity
          protein binding
          Rap guanyl-nucleotide exchange factor activity
          protein domain specific binding
          cAMP binding
          ion channel binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0