Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRMT5 SIRNA Assays ›
          Gene Symbol:
          PRMT5
          Gene Name:
          protein arginine methyltransferase 5
          Gene Aliases:
          HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs
          Chromosome Location:
          Chr.14: 22920511 - 22929585 on Build GRCh38
          UniGene ID:
          Hs.367854
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s20377
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 562.00
          Your Price
          Online offer: 533.65
          533.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRMT5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039619.2 NP_001034708.1 7 955
          NM_001282953.1 NP_001269882.1 6 823
          NM_001282954.1 NP_001269883.1 6 836
          NM_001282955.1 NP_001269884.1 6 738
          NM_006109.4 NP_006100.2 7 870
          XM_017020927.1 XP_016876416.1 6 594
          GenBank mRNA AB451246.1 - 7 646
          AB451370.1 - 7 646
          AF015913.1 - 7 646
          AF167572.1 - 7 737
          AK075251.1 - 7 748
          AK300863.1 - 6 620
          AK301812.1 - 6 629
          AK302240.1 - 6 518
          AK315500.1 - 7 686
          AL833612.1 - 7 655
          BC025979.2 - 7 658 3833019
          CR456741.1 - 7 646

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00272020_m1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal snRNP assembly
          DNA-templated transcription, termination
          regulation of transcription, DNA-templated
          regulation of mitotic nuclear division
          cell proliferation
          peptidyl-arginine methylation
          peptidyl-arginine methylation, to symmetrical-dimethyl arginine
          circadian regulation of gene expression
          peptidyl-arginine N-methylation
          endothelial cell activation
          histone H4-R3 methylation
          regulation of DNA methylation
          negative regulation of cell differentiation
          positive regulation of oligodendrocyte differentiation
          regulation of ERK1 and ERK2 cascade
          Golgi ribbon formation
          liver regeneration
          regulation of signal transduction by p53 class mediator
          negative regulation of nucleic acid-templated transcription
          positive regulation of adenylate cyclase-inhibiting dopamine receptor signaling pathway
          core promoter sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          methyltransferase activity
          methyl-CpG binding
          histone-arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          protein complex binding
          protein-arginine omega-N symmetric methyltransferase activity
          ribonucleoprotein complex binding
          histone methyltransferase activity (H4-R3 specific)
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0