Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GM13290 SIRNA Assays ›
          Gene Symbol:
          Gm13290 Gm13271 Gm13272 Gm13275 Gm13276 Gm13277 Gm13278 Gm13279 Gm13283 Gm13285 Gm13286 Gm13287 Gm13288 Gm13289 Ifnz
          Gene Name:
          predicted gene 13290
          predicted gene 13271
          predicted gene 13272
          predicted gene 13275
          predicted gene 13276
          predicted gene 13277
          predicted gene 13278
          predicted gene 13279
          predicted gene 13283
          predicted gene 13285
          predicted gene 13286
          predicted gene 13287
          predicted gene 13288
          predicted gene 13289
          interferon zeta
          Gene Aliases:
          OTTMUSG00000011291
          Ifnz, OTTMUSG00000011262
          OTTMUSG00000011265
          OTTMUSG00000011268
          OTTMUSG00000011269
          OTTMUSG00000011270
          OTTMUSG00000011272
          OTTMUSG00000011273
          EG545645, OTTMUSG00000011280
          OTTMUSG00000011282
          OTTMUSG00000011284
          OTTMUSG00000011285
          OTTMUSG00000011289
          OTTMUSG00000011290
          6030405N23Rik, BGIF, ENSMUSG00000073810, Lmtn
          Chromosome Location:
          Chr.4: 88773834 - 88774801 on Build GRCm38
          Chromosome Location:
          Chr.4: 88754868 - 88755416 on Build GRCm38
          Chromosome Location:
          Chr.4: 88779850 - 88780660 on Build GRCm38
          Chromosome Location:
          Chr.4: 88793742 - 88795297 on Build GRCm38
          Chromosome Location:
          Chr.4: 88785054 - 88786515 on Build GRCm38
          Chromosome Location:
          Chr.4: 88787977 - 88789438 on Build GRCm38
          Chromosome Location:
          Chr.4: 88790895 - 88792356 on Build GRCm38
          Chromosome Location:
          Chr.4: 88797237 - 88798204 on Build GRCm38
          Chromosome Location:
          Chr.4: 88760774 - 88761584 on Build GRCm38
          Chromosome Location:
          Chr.4: 88800320 - 88800868 on Build GRCm38
          Chromosome Location:
          Chr.4: 88757844 - 88758392 on Build GRCm38
          Chromosome Location:
          Chr.4: 88803254 - 88803802 on Build GRCm38
          Chromosome Location:
          Chr.4: 88805531 - 88808380 on Build GRCm38
          Chromosome Location:
          Chr.4: 88776762 - 88777729 on Build GRCm38
          Chromosome Location:
          Chr.4: 88782131 - 88783592 on Build GRCm38
          UniGene ID:
          Mm.380019
          Mm.473673
          Mm.368246
          Mm.457965
          Mm.474424
          Mm.473677
          Mm.149203
          Mm.489586
          Mm.489553
          Mm.478784
          Mm.489587
          Mm.380020
          Mm.487241
          Mm.380019
          Mm.368232
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s203805

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gm13290
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243155.1 NP_001230084.1 1 656
          GenBank mRNA AB024521.1 - 1 656
          Gene Symbol: Gm13271
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085528.1 NP_001078997.1 1 496
          Gene Symbol: Gm13272
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001161608.1 NP_001155080.1 1 496
          Gene Symbol: Gm13275
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085533.2 NP_001079002.1 1 1241
          Gene Symbol: Gm13276
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085532.3 NP_001079001.1 1 1151
          Gene Symbol: Gm13277
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098840.3 NP_001092310.1 1 1151
          Gene Symbol: Gm13278
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098841.3 NP_001092311.1 1 1151
          Gene Symbol: Gm13279
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243166.1 NP_001230095.1 1 656
          Gene Symbol: Gm13283
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085531.2 NP_001079000.1 1 496
          Gene Symbol: Gm13285
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001161609.1 NP_001155081.1 1 496
          Gene Symbol: Gm13286
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243150.1 NP_001230079.1 1 496
          Gene Symbol: Gm13287
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001161610.1 NP_001155082.1 1 496
          Gene Symbol: Gm13288
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243167.1 NP_001230096.1 1 1151
          GenBank mRNA AK031317.1 - 1 1152
          Gene Symbol: Gm13289
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243165.1 NP_001230094.1 656
          Gene Symbol: Ifnz
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_197889.2 NP_922871.1 1 1151
          GenBank mRNA AK046550.1 - 1 1152

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          biological_process
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          regulation of cell cycle
          apoptotic signaling pathway
          molecular_function
          cytokine activity
          type I interferon receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-c7vs9:80/100.66.73.137:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0