Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145568.3 | NP_663543.1 | 4 | 701 | ||
GenBank mRNA | AK050640.1 | - | 3 | 629 | ||
AK146818.1 | - | 3 | 640 | |||
AK148096.1 | - | 3 | 619 | |||
AK162678.1 | - | 3 | 629 | |||
BC005704.1 | - | 2 | 346 | 3598565 | ||
BC046798.1 | - | 3 | 597 | 6403577 | ||
BC069863.1 | - | 4 | 701 | 6306896 |