Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC099598.1 | - | 3 | 993 |
Gene Symbol: LOC100041034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK163333.1 | - | 4 | 1129 |