Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPIE SIRNA Assays ›
          Gene Symbol:
          PPIE
          Gene Name:
          peptidylprolyl isomerase E
          Gene Aliases:
          CYP-33, CYP33
          Chromosome Location:
          Chr.1: 39738845 - 39765698 on Build GRCh38
          UniGene ID:
          Hs.524690
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s20446
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPIE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195007.1 NP_001181936.1 6 356
          NM_001319293.1 NP_001306222.1 6 356
          NM_006112.3 NP_006103.1 6 356
          NM_203456.2 NP_982281.1 6 356
          NR_036543.1 6 348
          NR_036544.1 6 348
          XM_006710289.2 XP_006710352.1 6 311
          XM_006710290.3 XP_006710353.1 6 311
          XM_011540501.1 XP_011538803.1 6 458
          XM_017000051.1 XP_016855540.1 6 307
          XM_017000052.1 XP_016855541.1 6 458
          GenBank mRNA AF042385.1 - 6 360
          AF042386.1 - 6 360
          AF104012.1 - 6 311
          AF104013.1 - 6 311
          AK094660.1 - 5 1583
          AK130628.1 - 6 309
          AK293094.1 - 6 307
          AK304694.1 - 6 303
          AK313666.1 - 6 324
          AL833576.1 - 6 416
          BC004898.2 - 6 306 3545997
          BC008451.1 - 6 319 4134572
          BC107736.1 - 6 315 4809032
          BU160972.1 - 6 285
          BU179717.1 - 343
          BU196539.1 - 6 343
          CT002466.1 - 6 297
          DQ160195.1 - 6 300

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001047868, XM_006238796, XM_006238797, AY325152, BC158560.
          This siRNA also targets the following transcripts in Mouse: NM_019489, AK008177, BC045154, AF182825, AK042165.

          Matching Taqman Gene Expression Assay:


          Hs00237213_m1

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          protein peptidyl-prolyl isomerization
          transcription-coupled nucleotide-excision repair
          regulation of transcription, DNA-templated
          protein folding
          positive regulation of viral genome replication
          nucleotide binding
          RNA binding
          peptidyl-prolyl cis-trans isomerase activity
          protein binding
          cyclosporin A binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0