siRNA Details
Gene Symbol: Gm8350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087483.1 | - | 1, 2 | 140 |

More Information
Other Species Targeted


Gene Symbol: Gm8350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087483.1 | - | 1, 2 | 140 |