Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEMA4D SIRNA Assays ›
Gene Symbol:
SEMA4D
Gene Name:
semaphorin 4D
Gene Aliases:
C9orf164, CD100, M-sema-G, SEMAJ, coll-4
Chromosome Location:
Chr.9: 89360791 - 89498065 on Build GRCh38
UniGene ID:
Hs.494406
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20593
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEMA4D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142287.1
NP_001135759.1
9, 10
1077
NM_006378.3
NP_006369.3
9, 10
1077
XM_005251654.3
XP_005251711.1
7, 8
911
XM_011518123.2
XP_011516425.1
11, 12
1161
XM_011518124.2
XP_011516426.1
11, 12
1120
XM_011518125.1
XP_011516427.1
9, 10
1048
XM_011518127.2
XP_011516429.1
10, 11
1076
XM_011518128.2
XP_011516430.1
9, 10
1001
XM_011518129.1
XP_011516431.1
8, 9
930
XM_011518130.2
XP_011516432.1
8, 9
1165
XM_011518131.2
XP_011516433.1
7, 8
1103
XM_011518133.2
XP_011516435.1
8, 9
1027
XM_011518134.2
XP_011516436.1
6, 7
968
XM_017014193.1
XP_016869682.1
10, 11
2771
XM_017014194.1
XP_016869683.1
10, 11
1045
XM_017014195.1
XP_016869684.1
9, 10
982
XM_017014196.1
XP_016869685.1
9, 10
1319
XM_017014197.1
XP_016869686.1
8, 9
1203
XM_017014198.1
XP_016869687.1
7, 8
1140
XM_017014199.1
XP_016869688.1
11, 12
1120
GenBank mRNA
AB210030.1
-
7, 8
1282
AK293749.1
-
8, 10
1187
AK316107.1
-
9, 10
1271
BC054500.1
-
9, 10
1077
5551182
BX648216.1
-
7, 8
869
U60800.1
-
5, 6
592
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane-bound signaling molecule
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
positive regulation of protein phosphorylation
immune response
cell adhesion
negative regulation of cell adhesion
regulation of cell shape
negative regulation of alkaline phosphatase activity
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of cell migration
regulation of cell projection organization
negative regulation of apoptotic process
positive regulation of GTPase activity
ossification involved in bone maturation
negative regulation of osteoblast differentiation
positive regulation of collateral sprouting
regulation of dendrite morphogenesis
negative regulation of axon extension involved in axon guidance
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
leukocyte aggregation
semaphorin-plexin signaling pathway
semaphorin-plexin signaling pathway involved in bone trabecula morphogenesis
receptor activity
transmembrane signaling receptor activity
receptor binding
protein binding
semaphorin receptor binding
neuropilin binding
chemorepellent activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0