Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK2 SIRNA Assays ›
          Gene Symbol:
          CDK2
          Gene Name:
          cyclin dependent kinase 2
          Gene Aliases:
          CDKN2, p33(CDK2)
          Chromosome Location:
          Chr.12: 55966769 - 55972789 on Build GRCh38
          UniGene ID:
          Hs.19192
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s206
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290230.1 NP_001277159.1 2 423
          NM_001798.4 NP_001789.2 3 501
          NM_052827.3 NP_439892.2 3 501
          XM_011537732.1 XP_011536034.1 3 501
          GenBank mRNA AB012305.1 - 3 263
          AK291941.1 - 3 440
          AK293246.1 - 2 362
          BC003065.2 - 3 421 3504348
          BJ991087.1 - 3 498
          BT006821.1 - 3 261
          DA814453.1 - 3 501
          M68520.1 - 3 451
          X61622.1 - 3 261
          X62071.1 - 3 431

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00608082_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          G2/M transition of mitotic cell cycle
          lens development in camera-type eye
          DNA replication
          DNA repair
          potassium ion transport
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          mitotic nuclear division
          centriole replication
          meiotic nuclear division
          Ras protein signal transduction
          positive regulation of cell proliferation
          response to toxic substance
          histone phosphorylation
          peptidyl-serine phosphorylation
          organ regeneration
          mitotic G1 DNA damage checkpoint
          positive regulation of DNA-dependent DNA replication initiation
          response to estradiol
          cellular response to insulin stimulus
          response to drug
          response to ethanol
          positive regulation of transcription, DNA-templated
          response to cadmium ion
          centrosome duplication
          cell division
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          response to cAMP
          response to electrical stimulus
          regulation of gene silencing
          cellular response to nitric oxide
          regulation of signal transduction by p53 class mediator
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          cyclin binding
          protein complex binding
          histone kinase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0