Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081058.2 | NP_001074527.1 | 7, 8 | 3050 | ||
NM_027118.1 | NP_081394.1 | 7, 8 | 3050 | |||
GenBank mRNA | AK035493.1 | - | 7, 8 | 1721 | ||
AK173265.1 | - | 7, 8 | 3050 | |||
BC076619.1 | - | 8, 9 | 1505 |