Search
Search

Krt23| Gene Symbol: Krt23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033373.1 | NP_203537.1 | 3 | 659 | ||
| GenBank mRNA | AF102849.1 | - | 3 | 659 | ||
| AK036901.1 | - | 3 | 674 | |||
| AK036954.1 | - | 3 | 675 | |||
| AK145247.1 | - | 3 | 687 | |||
| BC003472.1 | - | 3 | 694 | |||
| BC019972.1 | - | 3 | 653 | 4187035 | ||