Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACADVL SIRNA Assays ›
          Gene Symbol:
          ACADVL
          Gene Name:
          acyl-CoA dehydrogenase, very long chain
          Gene Aliases:
          ACAD6, LCACD, VLCAD
          Chromosome Location:
          Chr.17: 7217125 - 7225267 on Build GRCh38
          UniGene ID:
          Hs.437178
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s207
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACADVL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000018.3 NP_000009.1 10 1170
          NM_001033859.2 NP_001029031.1 9 1104
          NM_001270447.1 NP_001257376.1 11 1106
          NM_001270448.1 NP_001257377.1 9 1245
          XM_006721516.2 XP_006721579.2 10 1123
          XM_011523829.1 XP_011522131.1 10 1123
          XM_011523830.1 XP_011522132.1 10 1123
          GenBank mRNA AK056083.1 - 9 1138
          AK056956.1 - 5 516
          AK058109.1 - 10 1052
          AK097243.1 - 8 1585
          AK222518.1 - 10 1052
          AK293549.1 - 11 1106
          BC000399.2 - 10 1108 2822593
          BC012912.1 - 10 1092 4471040
          BC020218.1 - 9 1012 5090104
          D43682.1 - 10 1049
          X86556.1 - 10 1101

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00817723_g1

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          fatty acid beta-oxidation
          energy derivation by oxidation of organic compounds
          epithelial cell differentiation
          fatty acid beta-oxidation using acyl-CoA dehydrogenase
          IRE1-mediated unfolded protein response
          very long-chain fatty acid catabolic process
          negative regulation of fatty acid biosynthetic process
          negative regulation of fatty acid oxidation
          lipid homeostasis
          regulation of cholesterol metabolic process
          fatty-acyl-CoA binding
          acyl-CoA dehydrogenase activity
          long-chain-acyl-CoA dehydrogenase activity
          electron carrier activity
          very-long-chain-acyl-CoA dehydrogenase activity
          flavin adenine dinucleotide binding
          oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0