Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC2 SIRNA Assays ›
Gene Symbol:
TACC2
Gene Name:
transforming acidic coiled-coil containing protein 2
Gene Aliases:
AZU-1, ECTACC
Chromosome Location:
Chr.10: 121989174 - 122254545 on Build GRCh38
UniGene ID:
Hs.501252
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20767
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291876.1
NP_001278805.1
11
7784
NM_001291877.1
NP_001278806.1
11
7931
NM_001291878.1
NP_001278807.1
7
2328
NM_001291879.1
NP_001278808.1
8
2142
NM_006997.3
NP_008928.1
6
2292
NM_206860.2
NP_996742.1
6
2292
NM_206861.2
NP_996743.1
9
2357
NM_206862.3
NP_996744.3
12
7919
XM_005269388.1
XP_005269445.1
13
7836
XM_005269389.4
XP_005269446.1
13
8360
XM_005269390.1
XP_005269447.1
14
7779
XM_005269391.1
XP_005269448.1
12
7776
XM_005269392.1
XP_005269449.1
13
7836
XM_005269393.1
XP_005269450.1
13
7743
XM_005269394.1
XP_005269451.1
12
7710
XM_005269395.1
XP_005269452.1
12
7701
XM_005269396.1
XP_005269453.1
13
7836
XM_005269397.1
XP_005269454.1
11
7575
XM_005269399.1
XP_005269456.1
12
7608
XM_006717548.1
XP_006717611.1
13
7731
XM_006717552.1
XP_006717615.1
14
7779
XM_006717553.1
XP_006717616.1
13
7743
XM_006717554.1
XP_006717617.1
13
7731
XM_011539126.1
XP_011537428.1
13
7833
XM_011539128.1
XP_011537430.1
11
2409
XM_011539129.1
XP_011537431.1
10
2274
XM_011539130.1
XP_011537432.1
10
2181
XM_011539131.1
XP_011537433.1
6
2247
XM_011539132.1
XP_011537434.1
7
2190
XM_011539133.1
XP_011537435.1
7
2095
XM_011539134.1
XP_011537436.1
7
2366
XM_017015484.1
XP_016870973.1
13
7824
XM_017015485.1
XP_016870974.1
14
7779
XM_017015486.1
XP_016870975.1
13
7743
XM_017015487.1
XP_016870976.1
13
7731
XM_017015488.1
XP_016870977.1
11
7470
XM_017015489.1
XP_016870978.1
11
7482
XM_017015491.1
XP_016870980.1
6
2387
XM_017015492.1
XP_016870981.1
6
2294
XM_017015493.1
XP_016870982.1
6
2142
XM_017015494.1
XP_016870983.1
6
2294
XM_017015495.1
XP_016870984.1
6
2154
XM_017015496.1
XP_016870985.1
7
2188
XM_017015497.1
XP_016870986.1
7
2447
XM_017015498.1
XP_016870987.1
6
2312
XM_017015499.1
XP_016870988.1
9
2371
GenBank mRNA
AF095791.2
-
6
1879
AF176646.1
-
7
2268
AF220152.2
-
4
1956
AF528098.1
-
9
2338
AF528099.1
-
12
7900
AK027066.1
-
4
1062
AK075097.1
-
4
420
AK295631.1
-
6
2075
AL713712.1
-
4
1368
AL833304.1
-
11
7018
BC039311.1
-
6
2292
BC141837.1
-
8
1980
BC142673.1
-
8
1980
BC144599.1
-
11
7899
9053129
BC144600.1
-
11
7752
9053130
BC150627.1
-
11
7752
9056997
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell proliferation
cerebral cortex development
nuclear hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0