Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAMKK2 SIRNA Assays ›
          Gene Symbol:
          CAMKK2
          Gene Name:
          calcium/calmodulin dependent protein kinase kinase 2
          Gene Aliases:
          CAMKK, CAMKKB
          Chromosome Location:
          Chr.12: 121237692 - 121298308 on Build GRCh38
          UniGene ID:
          Hs.297343
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s20926
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAMKK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270485.1 NP_001257414.1 9, 10 1079
          NM_001270486.1 NP_001257415.1 8, 9 963
          NM_006549.3 NP_006540.3 9, 10 1733
          NM_153499.2 NP_705719.2 9, 10 1733
          NM_153500.1 NP_705720.1 9, 10 1733
          NM_172214.2 NP_757363.1 9, 10 1733
          NM_172215.2 NP_757364.1 9, 10 1733
          NM_172216.1 NP_757365.1 9, 10 1733
          NM_172226.2 NP_757380.1 9, 10 1079
          XM_005253822.3 XP_005253879.1 9, 10 1035
          XM_005253823.1 XP_005253880.1 9, 10 1035
          XM_005253824.3 XP_005253881.1 9, 10 1035
          XM_011537763.1 XP_011536065.1 9, 10 1035
          XM_011537764.2 XP_011536066.1 9, 10 1081
          XM_017018694.1 XP_016874183.1 10, 11 1089
          XM_017018695.1 XP_016874184.1 10, 11 1298
          XM_017018696.1 XP_016874185.1 10, 11 1140
          XM_017018697.1 XP_016874186.1 10, 11 1162
          XM_017018698.1 XP_016874187.1 9, 10 1086
          XM_017018699.1 XP_016874188.1 10, 11 1089
          XM_017018700.1 XP_016874189.1 10, 11 1089
          XM_017018701.1 XP_016874190.1 9, 10 1086
          XM_017018702.1 XP_016874191.1 9, 10 1244
          XM_017018703.1 XP_016874192.1 10, 11 1089
          XM_017018704.1 XP_016874193.1 9, 10 1086
          XM_017018705.1 XP_016874194.1 10, 11 1089
          XM_017018706.1 XP_016874195.1 9, 10 1081
          XM_017018707.1 XP_016874196.1 8, 9 1004
          XM_017018708.1 XP_016874197.1 9, 10 1081
          XM_017018709.1 XP_016874198.1 8, 9 1001
          GenBank mRNA AB018330.2 - 8, 9 909
          AB081336.1 - 8, 9 936
          AB081337.1 - 8, 9 936
          AF101264.1 - 8, 9 556
          AF140507.1 - 9, 10 1017
          AF287630.1 - 9, 10 1727
          AF287631.1 - 9, 10 1727
          AF321385.1 - 9, 10 1483
          AF321386.1 - 9, 10 1483
          AF321387.1 - 9, 10 1483
          AF321388.1 - 9, 10 1483
          AK024748.1 - 8, 9 719
          AK292072.1 - 9, 10 1227
          AK300865.1 - 10, 11 979
          AL834322.1 - 3, 4 332
          BC000318.2 - 4, 5 208
          BC026060.2 - 9, 10 1012 3842706

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00198032_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein phosphorylation
          peptidyl-tyrosine phosphorylation
          calcium-mediated signaling
          regulation of protein kinase activity
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          calmodulin-dependent protein kinase activity
          protein tyrosine kinase activity
          calcium ion binding
          calmodulin binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mfr4m:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0