Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAMKK2 SIRNA Assays ›
          Gene Symbol:
          CAMKK2
          Gene Name:
          calcium/calmodulin dependent protein kinase kinase 2
          Gene Aliases:
          CAMKK, CAMKKB
          Chromosome Location:
          Chr.12: 121237692 - 121298308 on Build GRCh38
          UniGene ID:
          Hs.297343
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s20927
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAMKK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270485.1 NP_001257414.1 9, 10 1074
          NM_001270486.1 NP_001257415.1 8, 9 958
          NM_006549.3 NP_006540.3 9, 10 1728
          NM_153499.2 NP_705719.2 9, 10 1728
          NM_153500.1 NP_705720.1 9, 10 1728
          NM_172214.2 NP_757363.1 9, 10 1728
          NM_172215.2 NP_757364.1 9, 10 1728
          NM_172216.1 NP_757365.1 9, 10 1728
          NM_172226.2 NP_757380.1 9, 10 1074
          XM_005253822.3 XP_005253879.1 9, 10 1030
          XM_005253823.1 XP_005253880.1 9, 10 1030
          XM_005253824.3 XP_005253881.1 9, 10 1030
          XM_011537763.1 XP_011536065.1 9, 10 1030
          XM_011537764.2 XP_011536066.1 9, 10 1076
          XM_017018694.1 XP_016874183.1 10, 11 1084
          XM_017018695.1 XP_016874184.1 10, 11 1293
          XM_017018696.1 XP_016874185.1 10, 11 1135
          XM_017018697.1 XP_016874186.1 10, 11 1157
          XM_017018698.1 XP_016874187.1 9, 10 1081
          XM_017018699.1 XP_016874188.1 10, 11 1084
          XM_017018700.1 XP_016874189.1 10, 11 1084
          XM_017018701.1 XP_016874190.1 9, 10 1081
          XM_017018702.1 XP_016874191.1 9, 10 1239
          XM_017018703.1 XP_016874192.1 10, 11 1084
          XM_017018704.1 XP_016874193.1 9, 10 1081
          XM_017018705.1 XP_016874194.1 10, 11 1084
          XM_017018706.1 XP_016874195.1 9, 10 1076
          XM_017018707.1 XP_016874196.1 8, 9 999
          XM_017018708.1 XP_016874197.1 9, 10 1076
          XM_017018709.1 XP_016874198.1 8, 9 996
          GenBank mRNA AB018330.2 - 8, 9 904
          AB081336.1 - 8, 9 931
          AB081337.1 - 8, 9 931
          AF101264.1 - 8, 9 551
          AF140507.1 - 9, 10 1012
          AF287630.1 - 9, 10 1722
          AF287631.1 - 9, 10 1722
          AF321385.1 - 9, 10 1478
          AF321386.1 - 9, 10 1478
          AF321387.1 - 9, 10 1478
          AF321388.1 - 9, 10 1478
          AK024748.1 - 8, 9 714
          AK292072.1 - 9, 10 1222
          AK300865.1 - 10, 11 974
          AL834322.1 - 3, 4 327
          BC000318.2 - 4, 5 203
          BC026060.2 - 9, 10 1007 3842706

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00198032_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein phosphorylation
          peptidyl-tyrosine phosphorylation
          calcium-mediated signaling
          regulation of protein kinase activity
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          calmodulin-dependent protein kinase activity
          protein tyrosine kinase activity
          calcium ion binding
          calmodulin binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-d9lxt:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0