Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008509.2 | NP_032535.2 | 3 | 523 | ||
GenBank mRNA | AK002645.1 | - | 3 | 449 | ||
AK017272.1 | - | 521 | ||||
AK045064.1 | - | 3 | 412 | |||
AK079446.1 | - | 3 | 412 | |||
AK086023.1 | - | 3 | 344 | |||
AK150328.1 | - | 3 | 524 | |||
AK150355.1 | - | 3 | 524 | |||
AK150375.1 | - | 3 | 522 | |||
AK150457.1 | - | 3 | 419 | |||
AK150488.1 | - | 3 | 524 | |||
AK150505.1 | - | 3 | 524 | |||
AK150532.1 | - | 3 | 524 | |||
AK150577.1 | - | 3 | 524 | |||
AK150585.1 | - | 3 | 524 | |||
AK150593.1 | - | 3 | 524 | |||
AK150646.1 | - | 3 | 521 | |||
AK150672.1 | - | 3 | 442 | |||
AK150691.1 | - | 3 | 524 | |||
AK151006.1 | - | 3 | 524 | |||
AK151038.1 | - | 3 | 422 | |||
AK151049.1 | - | 3 | 524 | |||
AK151093.1 | - | 3 | 524 | |||
AK151099.1 | - | 3 | 524 | |||
AK151105.1 | - | 3 | 524 | |||
AK151112.1 | - | 3 | 525 | |||
AK151195.1 | - | 3 | 422 | |||
AK151216.1 | - | 3 | 525 | |||
AK151369.1 | - | 3 | 442 | |||
AK151434.1 | - | 3 | 442 | |||
AK151521.1 | - | 3 | 442 | |||
AK151540.1 | - | 3 | 412 | |||
AK151563.1 | - | 3 | 524 | |||
AK151630.1 | - | 3 | 422 | |||
AK151679.1 | - | 3 | 524 | |||
AK151698.1 | - | 3 | 524 | |||
AK151727.1 | - | 3 | 524 | |||
AK151772.1 | - | 3 | 524 | |||
AK151774.1 | - | 3 | 422 | |||
AK151801.1 | - | 3 | 412 | |||
AK151828.1 | - | 3 | 412 | |||
AK151866.1 | - | 3 | 524 | |||
AK151870.1 | - | 3 | 524 | |||
AK151872.1 | - | 3 | 524 | |||
AK151893.1 | - | 3 | 412 | |||
AK152014.1 | - | 3 | 524 | |||
AK152035.1 | - | 3 | 524 | |||
AK152049.1 | - | 3 | 524 | |||
AK152053.1 | - | 3 | 412 | |||
AK152079.1 | - | 3 | 524 | |||
AK152238.1 | - | 3 | 524 | |||
AK152251.1 | - | 3 | 521 | |||
AK152350.1 | - | 3 | 524 | |||
AK152390.1 | - | 3 | 363 | |||
AK152400.1 | - | 3 | 422 | |||
AK152657.1 | - | 3 | 524 | |||
AK152733.1 | - | 3 | 422 | |||
AK152775.1 | - | 3 | 524 | |||
AK152869.1 | - | 3 | 524 | |||
AK152956.1 | - | 3 | 524 | |||
AK153006.1 | - | 3 | 524 | |||
AK153148.1 | - | 3 | 524 | |||
AK153242.1 | - | 3 | 525 | |||
AK153425.1 | - | 3 | 524 | |||
AK153428.1 | - | 3 | 524 | |||
AK159268.1 | - | 3 | 524 | |||
AK170486.1 | - | 3 | 524 | |||
BC003305.1 | - | 3 | 512 | 3584788 | ||
BC158040.1 | - | 3 | 357 | 9088156 | ||
CT010344.1 | - | 3 | 325 | |||
J03302.1 | - | 3 | 301 | |||
M65258.1 | - | 3 | 485 |