Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172644.3 | NP_766232.1 | 11, 12 | 1641 | ||
GenBank mRNA | AK030952.1 | - | 11, 12 | 1630 | ||
AK033634.1 | - | 10, 11 | 2464 | |||
AK037337.1 | - | 11, 12 | 1660 | |||
AK048454.1 | - | 11, 12 | 1618 | |||
AK138809.1 | - | 11, 12 | 1642 | |||
AK154133.1 | - | 11, 12 | 1669 | |||
AK154348.1 | - | 11, 12 | 1669 | |||
AK164885.1 | - | 11, 12 | 1611 | |||
AK206226.1 | - | 1, 2 | 65 | |||
AK210088.1 | - | 2, 3 | 136 | |||
AK214891.1 | - | 1, 2 | 18 | |||
BC046429.1 | - | 4, 5 | 406 | |||
BC076606.1 | - | 11, 12 | 1633 | 30625515 | ||
BC089191.1 | - | 11, 12 | 1615 | 30473852 |