siRNA Details
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172644.3 | NP_766232.1 | 12 | 1664 | ||
GenBank mRNA | AK030952.1 | - | 12 | 1653 | ||
AK033634.1 | - | 11 | 2487 | |||
AK037337.1 | - | 12 | 1683 | |||
AK048454.1 | - | 12 | 1641 | |||
AK138809.1 | - | 12 | 1665 | |||
AK154133.1 | - | 12 | 1692 | |||
AK154348.1 | - | 12 | 1692 | |||
AK164885.1 | - | 12 | 1634 | |||
AK206226.1 | - | 2 | 88 | |||
AK210088.1 | - | 3 | 159 | |||
AK214891.1 | - | 2 | 41 | |||
BC046429.1 | - | 5 | 429 | |||
BC076606.1 | - | 12 | 1656 | 30625515 | ||
BC089191.1 | - | 12 | 1638 | 30473852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
